Learn More
Selleck Chemical LLC BMS-911172-E1628-25MG
SDP
Supplier:  Selleck Chemical LLC E162825MG
Encompass_Preferred
BMS-911172 is a brain penetrant selective inhibitor of adaptor protein 2-associated kinase 1 (AAK1) with an IC50 value of 12 nM BMS-911172 inhibits 2 phosphorylation and has potential as a therapeutic agent in neuropathic pain research
Catalog No. 50-313-7400