Learn More
Medchemexpress LLC Bms 986020 | 1257213-50-5 | MFCD30489743 | >=95.0% | 482.5 g/mol | C29H26N2O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10061950MG
Encompass_Preferred
BMS-986020 is a small-molecule, selective lysophosphatidic acid receptor 1 (LPA1) antagonist used as a research reagent in studies of pulmonary fibrosis and LPA1 signaling. It is supplied as a solid, white to yellow powder (CAS 1257213-50-5) with molecular formula C29H26N2O5 and molecular weight 482.53 g/mol.
  • Selective LPA1 antagonist suitable for receptor pharmacology studies.
  • Well-characterized chemical identity with published CAS, formula, and molecular weight.
  • Supplied as a solid powder appropriate for in vitro and preclinical assays.
  • Typical vendor-reported purity ≥95% (may vary by supplier; check lot certificate of analysis).
  • Recommended storage refrigerated or at -20 °C; follow supplier SDS for handling and safety.
Catalog No. 50-002-13665