Learn More
Medchemexpress LLC BMS-986121 | 313671-26-0 | 99.96% | C15H9Cl2N3O2S | 1 ML

Supplier: Medchemexpress LLC HY14151510MM/1ML
BMS-986121 is a positive allosteric modulator (PAM) of the μ opioid receptor, derived from patent WO2014107344. It is constructed on a chemical scaffold that represents a novel chemotype for μ receptor PAMs. For research use only.
- Positive allosteric modulator of the μ opioid receptor
- Augments β-arrestin-recruitment response
- Increases inhibition of forskolin-stimulated adenylyl cyclase activity
- Produces leftward shifts in the potency of endomorphin-I and leu-enkephalin
- Appears as a light yellow to yellow solid
- Store powder at -20°C for 3 years, 4°C for 2 years
- Store in solvent at -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.