Learn More
Medchemexpress LLC BMS-986141 | 1478711-48-6 | MFCD35066220 | C27H23N5O5S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY150790
Encompass_Preferred
BMS-986141 is an orally active, selective thrombin receptor protease-activated receptor-4 (PAR-4) antagonist with an IC50 value of 0.4 nM. It demonstrates excellent antithrombotic effects.
  • Orally active and selective PAR-4 antagonist
  • Demonstrates excellent antithrombotic effects
  • IC50 value of 0.4 nM for PAR-4
  • Inhibits platelet aggregation induced by PAR4 agonist peptide
  • Effective in animal models for anti-thrombotic activity
  • Involved in clinical trials for various thrombotic and cardiovascular conditions
Catalog No. 50-003-74618