Learn More
Selleck Chemical LLC BMS-986142
SDP
Supplier:  Selleck Chemical LLC S83811MG
Encompass_Preferred
BMS-986142 is a potent and highly selective reversible small molecule inhibitor of BTK with an IC50 of 0 5 nM In a panel of 384 kinases only five kinases were inhibited by BMS-986142 with less than 100-fold selectivity for BTK (TEC ITK BLK TXK and BMX)
Catalog No. 50-000-02890