Learn More
Medchemexpress LLC BMS-986158 | 1800340-40-2 | 99.8% | 495.62 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHY101567
Encompass_Preferred
BMS-986158 is a potent BET inhibitor effective in various cancer cell lines, with IC50s of 6.6 nM in NCI-H211 small cell lung cancer (SCLC) cells and 5 nM in MDA-MB231 triple negative breast cancer (TNBC) cells. It functions by inhibiting the bromodomain (BRD) and extra-terminal domain (BET) family of proteins, disrupting chromatin remodeling and preventing the expression of growth-promoting genes. This ultimately leads to an inhibition of tumor cell growth. It is intended for research use only.
  • Potent BET inhibitor
  • Inhibits bromodomain (BRD) and extra-terminal domain (BET) family proteins
  • Disrupts chromatin remodeling
  • Prevents expression of growth-promoting genes
  • Inhibits tumor cell growth
Catalog No. 50-003-64977