Learn More
Selleck Chemical LLC BMS-986158 S9691-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S969110MM/1ML
Encompass_Preferred
BMS-986158 is a potent inhibitor of BET with IC50s of 6 6 nM and 5 nM in NCI-H211 small cell lung cancer (SCLC) cells and MDA-MB231 triple negative breast cancer (TNBC) cells respectively
Catalog No. 50-000-24958