Learn More
Medchemexpress LLC BMS-986189 | 1629665-96-8 | 99.6% | 1887.21 | 50 MG

Supplier: Medchemexpress LLC HYP1037550MG
BMS-986189 is a macrocyclic peptide PD-1/PD-L1 interaction with an IC50 of 1.03 nM inhibitor. It can be used for cancer research, such as human lung carcinoma cells L2987.
- Appearance: Solid, white to off-white
- Purity: 99.58%
- Molecular weight: 1887.21
- Solubility in DMSO: ≥ 100 mg/mL
- Solubility in H2O: ≥ 1 mg/mL
- Storage (powder): -80°C for 2 years, -20°C for 1 year
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.