Learn More
Selleck Chemical LLC BMS-986235 E4628-25MG
SDP
Supplier:  Selleck Chemical LLC E462825MG
Encompass_Preferred
BMS-986235 (LAR-1219) is a selective orally active and potent agonist of formyl peptide receptor 2 (FPR2) with EC50 values of 0 41 nM and 3 4 nM for hFPR2 and mFPR2 respectively BMS-986235 therapy shows promising improvements in cardiac structure and function in a mouse model of heart failure highlighting its potential for preventing the progression of heart failure
Catalog No. 50-000-39104