Learn More
Medchemexpress LLC BMS-986238 | 2919596-13-5 | 99.9% | 2979.52 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1723205MG
Encompass_Preferred
BMS-986238 is an orally active macrocyclic peptide PD-L1 inhibitor. It can be used in the research of tumors such as solid tumors and lymphomas. PD-L1 is a key immune checkpoint protein that primarily regulates immune responses by inhibiting T-cell activity, serving as a core target for tumor immune escape and immunotherapy.
  • Orally active macrocyclic peptide PD-L1 inhibitor.
  • Applicable in the research of tumors including solid tumors and lymphomas.
  • Targets PD-L1, a key immune checkpoint protein, to inhibit T-cell activity.
Catalog No. 50-003-85361