Learn More
Medchemexpress LLC BMS-986238 | 2919596-13-5 | 99.9% | 2979.52 | C143H224N26O40S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17232010MG
Encompass_Preferred
BMS-986238 is a second-generation macrocyclic peptide designed as a potent inhibitor of programmed death-ligand 1 (PD-L1) for preclinical research. It is intended for use in studies of solid tumors and lymphomas and is supplied with supporting product and chemical information for laboratory applications.
  • Second-generation macrocyclic peptide scaffold suitable for research.
  • Potent PD-L1 inhibitory activity for immuno-oncology studies.
  • High reported purity (99.94%) supporting biochemical assays.
  • Available in small milligram pack sizes for compound screening.
  • Supplied with datasheet including CAS, molecular formula, and molecular weight.
Catalog No. 50-002-95393