Learn More
Medchemexpress LLC HY-139396 5mg , BMS-986299 CAS:2242952-69-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1393965MG
Encompass_Preferred
HY-139396 5mg BMS-986299 CAS: 2242952-69-6 BMS-986299 (compound 112) is a first-in-class NLRP3 inflammasome agonist with an EC50 of 1.28 μM. (patent WO2018152396A1). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1444