Learn More
TARGETMOL CHEMICALS INC BMS-P5 free base 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T22277L10MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. BMS-P5 free base is a specific and orally active peptidylarginine deiminase 4 (PAD4) inhibitor that blocks the formation of Neutrophil Extracellular Traps and delays the progression of multiple myeloma. Administration of BMS-P5 to multiple myeloma-bearing mice delays symptom onset and disease progression. Targeting PAD4 may be beneficial for treating multiple myeloma. Purity 99.96%
Catalog No. 50-287-7390