Learn More
Medchemexpress LLC HY-100608 5mg Medchemexpress, BMS453 CAS:166977-43-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1006085MG
Encompass_Preferred
Medchemexpress, HY-100608 5mg BMS453 CAS:166977-43-1 BMS453 (BMS-189453), a synthetic retinoid, is a RARβ agonist and a RARα/RARγ antagonist. BMS453 inhibits breast cell growth predominantly through the induction of active TGFβ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0596