Learn More
Cayman Chemical BMY 14802hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 422645 MG
Encompass_Preferred
An atypical antipsychotic; selectively binds to σ receptors over α1-ARs and dopamine D2 receptors (IC50s = 112, 520, and 6,340 nM, respectively); inhibits the conditioned avoidance response and apomorphine-induced stereotypy in rats (ED50s = 26.4 and 33 mg/kg, respectively)
Catalog No. 50-294-3403