Learn More
Selleck Chemical LLC BMY 7378 Dihydrochloride S2691-10mg
SDP
Supplier:  Selleck Chemical LLC S269110MG
Encompass_Preferred
BMY 7378 is a multi-targeted inhibitor of 2C-adrenoceptor and 1D-adrenoceptor with pKi of 6 54 and 8 2 respectively and acts as a mixed agonist and antagonist for 5-HT1A receptor with pKi of 8 3
Catalog No. 50-311-2559