Learn More
MSC ALPHATEC DISP COVERALL
Supplier:  MSC 00035840
Encompass
Disposable & Chemical Resistant Coveralls; Garment Style: Coveralls ; Size: Large ; Material: Polyethylene on Bicomponent; Polypropylene/Polyethylene Non-Woven ; Closure Type: 2-Way Zipper with Storm Flap ; Number Of Pockets: 0.000 ; Cuff Style: Elastic
Catalog No. 50-273-9470