Learn More
Medchemexpress LLC BNN6 | 106476-75-9 | 98.0% | 278.35 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1641175MG
Encompass_Preferred
BNN6 is a thermoresponsive nitric oxide (NO) donor. It functions by binding to a carrier and decomposes when stimulated by heat from photothermal action. This process releases high concentrations of nitric oxide, which exhibits anti-tumor effects by inducing apoptosis in tumor cells and inhibiting their repair. BNN6 can be used to create the multifunctional biosensor BNN6-BiTiS3-iRGD, which contributes to a synergistic anti-cancer effect when combined with photothermal therapy.
  • Works as a thermoresponsive nitric oxide donor.
  • Decomposes under heat stimulation to release nitric oxide.
  • Induces tumor cell apoptosis.
  • Inhibits tumor cell repair.
  • Can be used to synthesize a multifunctional biosensor for synergistic anti-cancer therapy with photothermal therapy.
Catalog No. 50-003-91219