Learn More
Medchemexpress LLC BnO-PEG5-OH | 57671-28-0 | 98.6% | 328.40 g/mol | C17H28O6 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW04271410G
Encompass_Preferred
BnO-PEG5-OH is a polyethylene glycol (PEG)-based PROTAC linker used to connect ligands in proteolysis targeting chimera (PROTAC) synthesis, providing spacer functionality and flexibility to aid target engagement and E3 ligase recruitment.
  • Polyethylene glycol spacer with five ethylene glycol units.
  • High purity suitable for research applications.
  • Liquid, colorless to light yellow, density ≈ 1.10 g/cm3.
  • Stable under recommended storage conditions for pure material and in solvent.
  • Available in common laboratory pack sizes, including 10 g.
Catalog No. 50-002-10587