Learn More
Medchemexpress LLC BnO-PEG6-OH | 24342-68-5 | 99.9% | 372.45 g/mol | C19H32O7 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW042654250MG
Encompass_Preferred
BnO-PEG6-OH is a benzyl-protected, hydroxyl-terminated polyethylene glycol linker containing six ethylene glycol units. It is supplied as a high-purity solid for use as a non-cleavable ADC linker and as a PEG-based linker in PROTAC synthesis.
  • High purity (99.88%) for reliable conjugation chemistry.
  • Six-unit PEG spacer provides flexibility and solubility.
  • Benzyl-protected hydroxyl end enables selective functionalization.
  • Available in multiple pack sizes including 250 mg for small-scale synthesis.
  • Stable when stored at -20°C (solid) and in solution at -80°C.
Catalog No. 50-002-02944