Learn More
Medchemexpress LLC BnO-PEG6-OH | 24342-68-5 | MFCD06797107 | 99.9% | 372.45 g/mol | C19H32O7 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW042654500MG
Encompass_Preferred
BnO-PEG6-OH is a non-cleavable six-unit polyethylene glycol (PEG) linker used for bioconjugation applications such as antibody-drug conjugates (ADCs) and proteolysis-targeting chimeras (PROTACs). It provides a flexible spacer to modulate steric and pharmacokinetic properties during conjugation workflows.
  • Non-cleavable six-unit PEG linker suitable for ADC and PROTAC synthesis.
  • High purity for research-grade applications.
  • Liquid, colorless to light yellow; density approximately 1.097 g/cm3.
  • Available in multiple package sizes, including 500 mg.
  • Stable when stored under recommended low-temperature conditions for extended periods.
Catalog No. 50-002-02860