Learn More
CPC Scientific H-Asp-Ser-Gly-Cys-Phe-Gly-Arg-Arg-Leu-Asp-Arg-Ile-Gly-Ser-Leu-Ser-Gly-Leu-Gly-Cys-Asn-Val-Leu-Arg-Arg-Tyr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NATR016B
Encompass
Endogenous neuropeptide with diuretic and hypotensive properties.ONE-LETTER SEQUENCE: DSGCFGRRLDRIGSLSGLGCNVLRRY (C4&C20 bridge)MOLECULAR FORMULA: C120H198N42O36S2MOLECULAR WEIGHT:2869.3STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [114547-28-3], SYNONYMS: BNP-26 (porcine), Brain Natriuretic Peptide-26 (porcine)RESEARCH AREA: CardiovascularREFERENCES: T Sudoh et. el Nature, 332(6159), 78-81 (1988)
Catalog No. 50-210-9871