Learn More
Abcam BNT x maleate, delta1 opioid receptor antagonist, 10MG
Supplier:  Abcam AB14614210MG
Encompass_Preferred
MW 545.6 Da, Purity >97%. Selective δ1 opioid receptor antagonist. Inhibits neurogenic ion transport in porcine ileal mucosa. Increases the antinociceptive ED50 value of DPDPE (ab120675) *in vivo.*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4932