Learn More
Medchemexpress LLC Bobcat339 hydrochloride | 2436747-44-1 | 99.6% | 334.20 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY111558A50MG
Encompass_Preferred
Bobcat339 hydrochloride is a potent and selective cytosine-based inhibitor of the TET enzyme, with IC50s of 33 μM and 73 μM for TET1 and TET2, respectively. It is useful in epigenetics and serves as a starting point for new therapeutics targeting DNA methylation and gene transcription.
  • Potent and selective cytosine-based inhibitor of TET enzyme.
  • Useful in epigenetics.
  • Serves as a starting point for new therapeutics that target DNA methylation and gene transcription.
Catalog No. 50-004-39933