Learn More
Selleck Chemical LLC Bobcat339 hydrochloride
SDP
Supplier:  Selleck Chemical LLC S66825MG
Encompass_Preferred
Bobcat339 is a selective cytosine-based TET enzyme inhibitor with IC50 of 33 M and 73 M for TET1 and TET2 respectively Bobcat339 can reduce DNA 5-hydroxymethylcytosine abundance by inhibiting TET enzyme function in living cells and provide support for its utility as a viable pharmacological probe
Catalog No. 50-000-01988