Learn More
Medchemexpress LLC Boc-11-aminoundecanoic acid | 10436-25-6 | 98.0% | 301.42 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0132535G
Encompass_Preferred
Boc-11-aminoundecanoic acid is an alkyl/ether-based PROTAC linker that can be utilized in the synthesis of MS432 (HY-130602). It is intended for research use only.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of MS432 (HY-130602)
Catalog No. 50-004-54768