Learn More
Medchemexpress LLC BOC-(1R,3S)-3-aminocyclopentane carboxylic acid | 161660-94-2 | 229.28 | 1 G

Supplier: Medchemexpress LLC HYW0048121G
BOC-(1R,3S)-3-aminocyclopentane carboxylic acid is a synthetic intermediate useful for pharmaceutical synthesis. It is intended for research use only.
- Useful for pharmaceutical synthesis.
- Store at room temperature for 3 years.
- In solvent, store at -80°C for 2 years or -20°C for 1 year.
- Soluble in DMSO at 100 mg/mL (436.15 mM), requiring ultrasonic for dissolution.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.