Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BOC-(1R,3S)-3-aminocyclopentane carboxylic acid | 161660-94-2 | 229.28 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYW004812

Encompass_Preferred

BOC-(1R,3S)-3-aminocyclopentane carboxylic acid is a synthetic intermediate useful for pharmaceutical synthesis.

  • Molecular formula: C11H19NO4
  • Appearance: Solid
  • Color: White to off-white
  • Solubility (in vitro): DMSO 100 mg/mL (requires ultrasonic)
  • Storage: Room temperature for 3 years; in solvent: -80°C for 2 years or -20°C for 1 year

Catalog No. 50-004-47026


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.