Learn More
Medchemexpress LLC BOC-(1S,3R)-3-Aminocyclopentanecarboxylic acid | 261165-05-3 | 229.27 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW006202
Encompass_Preferred
BOC-(1S,3R)-3-aminocyclopentanecarboxylic acid is a synthetic intermediate useful for pharmaceutical synthesis. This product is for research use only and not sold to patients.
  • Synthetic intermediate useful for pharmaceutical synthesis
  • Available in solid form, white to off-white color
  • Can be stored at room temperature for 3 years
  • In solvent, stored at -80°C for 2 years, and -20°C for 1 year
Catalog No. 50-004-48935