Learn More
Medchemexpress LLC Boc-2-Nal-Oh | 58438-04-3 | 98.84% | 315.36 | 25 G

Supplier: Medchemexpress LLC 25GHYW011321
Boc-2-Nal-OH is an alanine derivative. Amino acids and amino acid derivatives are commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, fuel supply during exercise, mental performance during stress-related tasks, and help prevent exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
- Appearance: Solid
- Color: White to off-white
- Shipping: Room temperature in continental US
- Storage powder: -20°C for 3 years, 4°C for 2 years
- Storage in solvent: -80°C for 6 months, -20°C for 1 month
- Soluble in ethanol
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.