Learn More
Medchemexpress LLC Boc-6-aminohexanoic acid | 6404-29-1 | 231.29 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00752910G
Encompass_Preferred
Boc-6-aminohexanoic acid is an alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. It is suitable for use as a building block in various chemical syntheses.
  • Alkyl/ether-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Appears as a white to off-white solid
  • Soluble in H2O: 50 mg/mL
Catalog No. 50-004-46347