Learn More
Medchemexpress LLC Boc-6-aminohexanoic acid | 6404-29-1 | 231.29 | 25 G

Supplier: Medchemexpress LLC 25GHYW007529
Boc-6-aminohexanoic acid is an alkyl/ether-based PROTAC linker designed for the synthesis of PROTACs. These compounds exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by linking a ligand for an E3 ubiquitin ligase with a ligand for the target protein. This product is for research use only.
- Alkyl/ether-based PROTAC linker
- Used in the synthesis of PROTACs
- Exploits intracellular ubiquitin-proteasome system
- Selectively degrades target proteins
- Purity of 98.0%
- Molecular weight of 231.29
- Appearance as a white to off-white solid
- Soluble in H2O at 50 mg/mL (requires ultrasonic)
- Stable as powder at -20°C for 3 years, or 4°C for 2 years
- Stable in solvent at -80°C for 6 months, or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.