Learn More
Medchemexpress LLC Boc-Ala-Ala-Asp-pNA | 201732-83-4 | 99.1% | 495.48 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP44535MG
Encompass_Preferred
Boc-Ala-Ala-Asp-pNA is a chromogenic substrate of granzyme B, utilized for testing the functional activity of granzyme B. This product is for research purposes only and is not intended for medical, clinical diagnostic, or food use.
  • Chromogenic substrate
  • Tests granzyme B functional activity
  • High purity: 99.1%
  • Stable under sealed storage, away from moisture
  • Available in various quantities
  • Solid, white to off-white appearance
Catalog No. 50-003-97678