Learn More
Medchemexpress LLC Boc-Ala-OMe | 28875-17-4 | MFCD00038513 | 203.24 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW002173500G
Encompass_Preferred
Boc-Ala-OMe is an alanine derivative primarily used in research. This amino acid derivative can act as an ergogenic supplement, influencing anabolic hormone secretion and fuel supply during exercise.
  • Influences anabolic hormone secretion
  • Supplies fuel during exercise
  • Improves mental performance during stress-related tasks
  • Helps prevent exercise-induced muscle damage
  • Soluble in DMSO (100 mg/mL)
  • Stable under recommended storage conditions
Catalog No. 50-004-47159