Learn More
Medchemexpress LLC Boc-amido-PEG9-amine | 890091-43-7 | ≥98.0% | C25H52N2O11 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140195
Encompass_Preferred
Boc-amido-PEG9-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs operate by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Not for sale to patients
Catalog No. 50-003-52561