Learn More
Medchemexpress LLC Boc-aminooxy-amide-PEG4-propargyl | 2253965-01-2 | 404.46 g/mol | C18H32N2O8 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY133436250MG
Encompass_Preferred
Boc-aminooxy-amide-PEG4-propargyl is a Boc-protected aminooxy-containing 4-unit PEG linker used as a non-cleavable ADC linker and for bioconjugation. It provides an aminooxy functionality for oxime ligation and a propargyl (alkyne) handle for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Non-cleavable 4-unit PEG linker
  • Provides aminooxy functionality for oxime ligation
  • Provides propargyl (alkyne) handle for click chemistry
  • Suitable for ADC linker synthesis and bioconjugation
  • Available in multiple pack sizes for synthesis workflows
Catalog No. 50-002-02979