Learn More
Medchemexpress LLC Boc-Aminooxy-PEG4-NH2 | 2062663-66-3 | 95.0% | C13H28N2O7 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140438
Encompass_Preferred
Boc-Aminooxy-PEG4-NH2 is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules are instrumental in drug discovery, facilitating the targeted degradation of specific proteins. By leveraging the intracellular ubiquitin-proteasome system, PROTACs offer a novel approach to modulating protein function.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Facilitates targeted protein degradation
  • Utilizes the ubiquitin-proteasome system
Catalog No. 50-003-52770