Learn More
Medchemexpress LLC Boc-Aminooxy-PEG4-Tos | 1807539-01-0 | C20H33NO9S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140377100MG
Encompass_Preferred
Boc-Aminooxy-PEG4-Tos is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed to degrade target proteins by exploiting the intracellular ubiquitin-proteasome system. They consist of two different ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other targets the protein intended for degradation. This product is for research use only.
  • Functions as a PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Contributes to targeted protein degradation.
  • Should be stored at -20°C for up to 3 years in pure form, or at 4°C for up to 2 years in pure form.
  • When in solvent, store at -80°C for up to 6 months, or at -20°C for up to 1 month.
  • Appears as a colorless to light yellow viscous liquid.
Catalog No. 50-003-56191