Learn More
Medchemexpress LLC Boc-Aminooxy-PEG4-Tos | 1807539-01-0 | C20H33NO9S | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140377
Encompass_Preferred
Boc-Aminooxy-PEG4-Tos is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs are innovative molecules that harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins by linking a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system for selective protein degradation
Catalog No. 50-003-52647