Learn More
Medchemexpress LLC Boc-Arg(di-Z)-OH | 51219-19-3 | 542.58 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW013097
Encompass_Preferred
Boc-Arg(di-Z)-OH can be used for the synthesis of amino acid. It can also be used for the research of inhibitors for processing proteinases and is coupled via the mixed anhydride (MA) with HGlu(OBzl)-Lys(Z)-Arg(Z,Z)-CH2Cl.
  • Used for the synthesis of amino acid.
  • Used for the research of inhibitors for processing proteinases.
Catalog No. 50-004-51408