Learn More
Medchemexpress LLC Boc-Arg-Val-Arg-Arg-AMC hydrochloride | 136132-77-9 | 99.5% | 879.45 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP454810MG
Encompass_Preferred
Boc-Arg-Val-Arg-Arg-AMC hydrochloride (Boc-RVRR-AMC) is a synthetic fluorogenic substrate efficiently cleaved by furin, making it a valuable tool for research applications. This product is provided as a solid, typically white to off-white in color, and requires sealed storage away from moisture and light to maintain its stability. It is soluble in water for in vitro applications.
  • Synthetic fluorogenic substrate
  • Efficiently cleaved by furin
  • Available as a white to off-white solid
  • Requires sealed storage away from moisture and light
  • Soluble in water
Catalog No. 50-003-98824