Learn More
Medchemexpress LLC Boc-Asp(OMe)-fluoromethyl ketone | 187389-53-3 | MFCD03453073 | 98.0% | 263.26 g·mol⁻¹ | C11H18FNO5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10334810MM 1ML
Encompass_Preferred
Boc-Asp(OMe)-fluoromethyl ketone is a cell-permeable, broad-spectrum caspase inhibitor used in vitro to block caspase-mediated apoptosis and Fas-mediated processes. It is supplied as ready-to-use solutions and as milligram quantities for research applications.
  • Broad-spectrum caspase inhibitor suitable for apoptosis and cell-death studies.
  • Does not affect IL-8 chemotactic activity in reported assays.
  • Available as a 10 mM solution in DMSO and in multiple mg quantities for flexibility in experimental design.
  • High stated purity (98.0%) for reliable activity in assays.
  • Molecular formula C11H18FNO5 and molecular weight 263.26 g·mol⁻¹.
Catalog No. 50-002-45056