Learn More
Medchemexpress LLC Boc-Asp(OMe)-fluoromethyl ketone | 187389-53-3 | MFCD03453073 | 98.0% | 263.26 g·mol⁻¹ | C11H18FNO5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10334825MG
Encompass_Preferred
Boc-Asp(OMe)-fluoromethyl ketone is a cell-permeable, broad-spectrum caspase inhibitor supplied for research use. It inhibits Fas-mediated phagocytosis and oxidative rupture while not affecting IL-8-mediated chemotaxis. Typical applications include caspase activity assays and studies of apoptosis signaling.
  • Broad-spectrum caspase inhibitor for biochemical and cell-based assays.
  • Cell-permeable small molecule suitable for intracellular studies.
  • High purity (98.0%) for reliable experimental results.
  • Available in multiple pack sizes and as a DMSO solution for convenience.
  • Supplied for research use only; follow the SDS for handling and storage.
Catalog No. 50-002-44762