Learn More
Medchemexpress LLC Boc-Asp(OMe)-fluoromethyl ketone | 187389-53-3 | MFCD03453073 | 98.0% | 263.26 g/mol | C11H18FNO5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1033485MG
Encompass_Preferred
Boc-Asp(OMe)-fluoromethyl ketone is a fluoromethyl ketone peptide that irreversibly inhibits caspases and is used to block caspase-dependent apoptosis in biochemical and cell-based assays.
  • Broad-range caspase inhibitor, effective in blocking apoptosis-related protease activity.
  • Inhibits Fas-mediated phagocytosis and oxidative rupture without affecting IL-8 chemotaxis.
  • Suitable for caspase activity assays and apoptosis research in vitro and in cell-based studies.
  • High purity (≈98.0%) and supplied as a colorless to light yellow liquid.
  • Soluble in DMSO at concentrations ≥100 mg/mL for stock solutions.
  • Store sealed, away from moisture at -20°C; in solution keep at -80°C for long-term storage.
Catalog No. 50-002-45055