Learn More
Medchemexpress LLC Boc-Asp(OMe)-fluoromethyl ketone | 187389-53-3 | MFCD03453073 | 98.0% | 263.26 g/mol | C11H18FNO5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10334850MG
Encompass_Preferred
Boc-Asp(OMe)-fluoromethyl ketone is a cell-permeable, broad-spectrum caspase inhibitor used in research to block caspase-dependent processes such as Fas-mediated phagocytosis and oxidative rupture while not affecting IL-8 chemotaxis.
  • Broad-spectrum caspase inhibitor for biochemical and cell-based studies.
  • Cell-permeable fluoromethyl ketone scaffold.
  • High purity (98.0%).
  • Molecular formula C11H18FNO5; molecular weight 263.26 g/mol.
  • Available in multiple pack sizes, including a 50 MG vial.
Catalog No. 50-002-44726