Learn More
Medchemexpress LLC Boc-C1-PEG2-C4-Cl (PROTAC Linker 1) | 1835705-53-7 | 99.8% | 294.81 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10837150MG
Encompass_Preferred
Boc-C1-PEG2-C4-Cl (PROTAC Linker 1) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This technology utilizes small molecules to recruit target proteins for ubiquitination and removal by the proteasome. It can mediate the degradation of c-ABL and BCR-ABL by recruiting Cereblon or Von Hippel Lindau E3 ligases.
  • Appearance: liquid (density: 1.026±0.06 g/cm³)
  • Color: colorless to light yellow
  • Used in the synthesis of PROTACs
  • Facilitates the degradation of oncogenic BCR-ABL
Catalog No. 50-003-47152