Learn More
Medchemexpress LLC Boc-C2-NH2 | 15231-41-1 | MFCD06212562 | 99.8% | 145.20 g/mol | C7H15NO2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW017970500MG
Encompass_Preferred
Boc-C2-NH2 is a Boc-protected C2 alkyl/ether-based PROTAC linker (tert-butyl 3-aminopropanoate) used in medicinal chemistry to introduce a protected amino-terminated two-carbon spacer during synthesis of bifunctional degraders. It is supplied as a colorless to light yellow liquid with high reported purity and is intended for incorporation into linker architectures where controlled deprotection and subsequent coupling are required.
  • Boc-protected amino terminus for controlled deprotection
  • Short two-carbon spacer suitable for compact linker design
  • High reported purity reduces downstream purification
  • Compatible with standard amide coupling and deprotection chemistries
  • Available in multiple laboratory-scale pack sizes
Catalog No. 50-003-01402