Learn More
Medchemexpress LLC Boc-C5-O-C5-O-C6-Cl | 1835705-52-6 | 393.00 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10837210MM/1ML
Encompass_Preferred
Boc-C5-O-C5-O-C6-Cl (PROTAC Linker 2) is a PROTAC linker utilized to connect the respective tyrosine kinase inhibitor (TKI) to the E3 recruiting ligand. It is for research use only.
  • PROTAC linker utilized to connect the respective tyrosine kinase inhibitor (TKI) to the E3 recruiting ligand.
  • Alkyl/ether linker type.
Catalog No. 50-003-47153