Learn More
Medchemexpress LLC Boc-Cys(Npys)-OH | 76880-29-0 | MFCD00153301 | 98.0% | 375.42 g/mol | C13H17N3O6S2 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1417791G
Encompass_Preferred
Boc-Cys(Npys)-OH is a Boc-protected cysteine derivative in which the cysteine thiol is protected with an Npys (3-nitro-2-pyridylsulfanyl) group. It is supplied as a light yellow solid for use in peptide synthesis and thiol-directed chemistry, facilitating selective thiol manipulation and site-directed disulfide bond formation.
  • Boc-protected amine suitable for standard peptide coupling
  • Npys thiol protecting group enables directed disulfide formation
  • Suitable for solid-phase and solution-phase peptide synthesis
  • High purity for consistent synthetic results
  • Light yellow solid, easy to handle and store
Catalog No. 50-002-83446