Learn More
Bachem Boc-Cys(SEt)-OH · DCHA
SDP
Supplier:  Bachem 4000384.0001
Encompass_Preferred
Boc-Cys(SEt)-OH · DCHA, 25461-01-2, C10H19NO4S2 · C12H23N, Mr 462.72, 1g. The ethylthio group is cleaved by mercaptans.
Catalog No. 50-312-139